missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl chloride, 95%, stab. with sodium carbonate, Thermo Scientific Chemicals

Product Code. 11447027
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cinnamyl chloride is used in the enantioselective total synthesis of helioporins C and E, bioactive marine diterpenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cinnamyl chloride is used in the enantioselective total synthesis of helioporins C and E, bioactive marine diterpenes.

Solubility
Soluble in water (0.2 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2687-12-9

Specifications

Melting Point 7°C
Density 1.096
Boiling Point 86°C to 87°C (2 mmHg)
Flash Point 79°C (174°F)
Molecular Formula C9H9Cl
Refractive Index 1.5845
MDL Number MFCD00000986
Quantity 1000 g
UN Number UN2927
Beilstein 741974
Synonym cinnamyl chloride, 3-chloroprop-1-en-1-yl benzene, 3-chloro-1-phenyl-1-propene, trans-cinnamyl chloride, 1e-3-chloroprop-1-en-1-yl benzene, benzene, 3-chloropropenyl, e-3-chloroprop-1-enyl benzene, unii-2i1adl56tx, 2i1adl56tx, cinnamylchloride
Solubility Information Soluble in water (0.2g/L at 20°C).
InChI Key IWTYTFSSTWXZFU-QPJJXVBHSA-N
SMILES C1=CC=C(C=C1)C=CCCl
IUPAC Name [(E)-3-chloroprop-1-enyl]benzene
Molecular Weight (g/mol) 152.62
PubChem CID 639658
Formula Weight 152.62
Percent Purity 95%
Chemical Name or Material Cinnamyl chloride, stabilized with sodium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'