missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl chloride, 95% trans, Thermo Scientific Chemicals

Product Code. 10674872
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2687-12-9
Molecular Formula C9H9Cl
Molecular Weight (g/mol) 152.62
MDL Number MFCD00000986
InChI Key IWTYTFSSTWXZFU-QPJJXVBHSA-N
Synonym cinnamyl chloride, 3-chloroprop-1-en-1-yl benzene, 3-chloro-1-phenyl-1-propene, trans-cinnamyl chloride, 1e-3-chloroprop-1-en-1-yl benzene, benzene, 3-chloropropenyl, e-3-chloroprop-1-enyl benzene, unii-2i1adl56tx, 2i1adl56tx, cinnamylchloride
PubChem CID 639658
IUPAC Name [(E)-3-chloroprop-1-enyl]benzene
SMILES C1=CC=C(C=C1)C=CCCl

Specifications

Melting Point 7.0°C
Color Brown-Yellow to Yellow
Density 1.0960g/mL
Boiling Point 120.0°C to 126.0°C (15.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5CH=CHCH2Cl
Refractive Index 1.5830 to 1.5850
Quantity 5 g
Beilstein 05, II, 372
Fieser 13,299
Specific Gravity 1.096
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, n,n-dmf,, acetone, formic acid, benzene, ethyl ether
Formula Weight 152.62
Percent Purity 95% trans
Physical Form Liquid
Chemical Name or Material Cinnamyl chloride, 95% trans
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cinnamyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'