Learn More
Thermo Scientific Chemicals Ciprofloxacin, 98%
Ciprofloxacin, CAS# 85721-33-1, is a second-generation fluoroquinolone showing antibiotic activity commonly used against susceptible organisms mainly gram-negative bacteria.
£28.34 - £80.70
Chemical Identifiers
CAS | 85721-33-1 |
---|---|
Molecular Formula | C17H18FN3O3 |
Molecular Weight (g/mol) | 331.34 |
InChI Key | MYSWGUAQZAJSOK-UHFFFAOYSA-N |
Synonym | ciprofloxacin, ciprofloxacine, ciprobay, ciproxan, ciprofloxacina, cipro, ciprofloxacino, ciprofloxacinum, cipro iv, bacquinor |
PubChem CID | 2764 |
ChEBI | CHEBI:100241 |
IUPAC Name | 1-cyclopropyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid |
SMILES | C1CC1N2C=C(C(=O)C3=CC(=C(C=C32)N4CCNCC4)F)C(=O)O |
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
13531640
|
Thermo Scientific Chemicals
449620050 |
5 g | Glass Bottle |
£28.34
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13541640
|
Thermo Scientific Chemicals
449620250 |
25 g | Glass Bottle |
£80.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• Ciprofloxacin is considered a synthetic broad-spectrum second-generation compound with antibiotic activity mainly against gram-negative bacteria
• It inhibits bacterial DNA gyrase and topoisomerase IV, enzymes that are essential for DNA replication, cell division, and causes double-strand breaks in the bacterial chromosome
Applications
• Ciprofloxacin presents more activity against gram-negative bacteria than gram-positive bacteria.
• Ciprofloxacin does not present cross-resistance between fluoroquinolones and other antibiotics classes, which is beneficial in laboratory experiments when other antibiotics are no longer effective
Chemische Identifikatoren
85721-33-1 | |
331.34 | |
ciprofloxacin, ciprofloxacine, ciprobay, ciproxan, ciprofloxacina, cipro, ciprofloxacino, ciprofloxacinum, cipro iv, bacquinor | |
CHEBI:100241 | |
C1CC1N2C=C(C(=O)C3=CC(=C(C=C32)N4CCNCC4)F)C(=O)O |
C17H18FN3O3 | |
MYSWGUAQZAJSOK-UHFFFAOYSA-N | |
2764 | |
1-cyclopropyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid |
Spezifikation
85721-33-1 | |
C17H18FN3O3 | |
ciprofloxacin, ciprofloxacine, ciprobay, ciproxan, ciprofloxacina, cipro, ciprofloxacino, ciprofloxacinum, cipro iv, bacquinor | |
MYSWGUAQZAJSOK-UHFFFAOYSA-N | |
1-cyclopropyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid | |
2764 | |
331.34 | |
Ciprofloxacin |
97.5% min. (HPLC) | |
15,2313 | |
Solubility in water: insoluble. Other solubilities: insoluble in ethanol, soluble in hot dmso,soluble in hot methanol | |
C1CC1N2C=C(C(=O)C3=CC(=C(C=C32)N4CCNCC4)F)C(=O)O | |
331.34 | |
CHEBI:100241 | |
98% |
Sicherheit und Handhabung
- Ciprofloxacin
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.