missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Cyclopentanediol, 98%, Thermo Scientific Chemicals

£39.09 - £117.59

Chemical Identifiers

CAS 5057-98-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00134192
InChI Key VCVOSERVUCJNPR-SYDPRGILSA-N
Synonym cis-1,2-cyclopentanediol, cis-cyclopentane-1,2-diol, cis-1,2-dihydroxycyclopentane, 1r,2s-cyclopentane-1,2-diol, 1s,2r cyclopentane-1,2-diol, cis-1,2-cyclopentandiol, 1,2-cis-cyclopentanediol, cis-1,2-cyclopen-tanediol, cis-cyclopenten-1,2-diol, 1alpha,2alpha-cyclopentanediol
PubChem CID 2733293
IUPAC Name (1R,2S)-cyclopentane-1,2-diol
SMILES C1CC(C(C1)O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10013750
View Documents Promotion Details
Thermo Scientific Acros
310290010
1 g Glass bottle
£39.09
1g
Please to purchase this item. Need a web account? Register with us today!
10444935
View Documents Promotion Details
Thermo Scientific Acros
310290050
5 g Glass bottle
£117.59
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5057-98-7
102.13
VCVOSERVUCJNPR-SYDPRGILSA-N
2733293
C1CC(C(C1)O)O
C5H10O2
MFCD00134192
cis-1,2-cyclopentanediol, cis-cyclopentane-1,2-diol, cis-1,2-dihydroxycyclopentane, 1r,2s-cyclopentane-1,2-diol, 1s,2r cyclopentane-1,2-diol, cis-1,2-cyclopentandiol, 1,2-cis-cyclopentanediol, cis-1,2-cyclopen-tanediol, cis-cyclopenten-1,2-diol, 1alpha,2alpha-cyclopentanediol
(1R,2S)-cyclopentane-1,2-diol

Specifications

5057-98-7
Yellow
>110°C
97.5% min. (GC)
C5H8(OH)2
MFCD00134192
Solubility in water: partially soluble in water.
C1CC(C(C1)O)O
102.13
102.13
Liquid After Melting
32.0°C to 33.0°C
83.0°C to 83.5°C (5.0 mmHg)
Authentic
C5H10O2
1.4770 to 1.4790
cis-1,2-cyclopentanediol, cis-cyclopentane-1,2-diol, cis-1,2-dihydroxycyclopentane, 1r,2s-cyclopentane-1,2-diol, 1s,2r cyclopentane-1,2-diol, cis-1,2-cyclopentandiol, 1,2-cis-cyclopentanediol, cis-1,2-cyclopen-tanediol, cis-cyclopenten-1,2-diol, 1alpha,2alpha-cyclopentanediol
VCVOSERVUCJNPR-SYDPRGILSA-N
(1R,2S)-cyclopentane-1,2-diol
2733293
98%
cis-1,2-Cyclopentanediol
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only