Learn More
cis-2-Butene-1,4-diol, 97%, Thermo Scientific Chemicals
£19.90 - £252.26
Chemical Identifiers
CAS | 6117-80-2 |
---|---|
Molecular Formula | C4H8O2 |
Molecular Weight (g/mol) | 88.11 |
MDL Number | MFCD00002924,MFCD00063207 |
InChI Key | ORTVZLZNOYNASJ-UPHRSURJSA-N |
Synonym | cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv |
PubChem CID | 643790 |
SMILES | OC\C=C/CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10085753
|
Thermo Scientific Acros
107721000 |
100 mL | Glass bottle |
£19.90
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10254610
|
Thermo Scientific Acros
107725000 |
500 mL | Glass bottle |
£56.38
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11954631
|
Thermo Scientific Acros
107720025 |
2.5 L | Glass bottle |
£252.26
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6117-80-2 | |
88.11 | |
ORTVZLZNOYNASJ-UPHRSURJSA-N | |
643790 |
C4H8O2 | |
MFCD00002924,MFCD00063207 | |
cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv | |
OC\C=C/CO |
Specifications
6117-80-2 | |
Colorless | |
235.0°C | |
Authentic | |
C4H8O2 | |
1.4770 to 1.4800 | |
1.07 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone,slightly soluble in benzene | |
OC\C=C/CO | |
88.11 | |
88.11 | |
Liquid |
7.0°C | |
1.0700g/mL | |
128°C | |
96% min. (cis-isomer) (GC) | |
HOCH2CH=CHCH2OH | |
MFCD00002924,MFCD00063207 | |
cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv | |
ORTVZLZNOYNASJ-UPHRSURJSA-N | |
(Z)-but-2-ene-1,4-diol | |
643790 | |
97% | |
cis-2-Butene-1, 4-diol, 97% cis |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 228-085-1
Recommended Storage : below 6°C, crystals may form, agitation necessary to disperse
- cis-2-Butene-1,4-diol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.