missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-3-Hepten-1-ol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
cis-3-Hepten-1-ol is used in the analysis of hop-derived terpenoids in beer. Imparts a fresh flower garden odor so used in flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-3-Hepten-1-ol is used in the analysis of hop-derived terpenoids in beer. Imparts a fresh flower garden odor so used in flavoring agent.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
cis-3-Hepten-1-ol is used in the analysis of hop-derived terpenoids in beer. Imparts a fresh flower garden odor so used in flavoring agent.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.85 |
Boiling Point | 92°C (60 mmHg) |
Flash Point | 68°C (154°F) |
Refractive Index | 1.444 |
Quantity | 5 g |
Beilstein | 1719803 |
Solubility Information | Insoluble in water. |
Formula Weight | 114.18 |
Percent Purity | 97% |
Chemical Name or Material | cis-3-Hepten-1-ol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'