missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Hexen-1-ol, 98%, Thermo Scientific Chemicals

Product Code. 11434073
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-96-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00063217
InChI Key UFLHIIWVXFIJGU-ARJAWSKDSA-N
Synonym z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z
PubChem CID 5281167
ChEBI CHEBI:28857
SMILES CC\C=C/CCO

Specifications

Density 0.848
Boiling Point 156°C to 157°C
Flash Point 44°C (111°F)
Odor Pleasant
Refractive Index 1.44
Quantity 50 g
UN Number UN1987
Beilstein 1719712
Merck Index 14,4700
IUPAC Name (Z)-hex-3-en-1-ol
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material cis-3-Hexen-1-ol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • cis-3-Hexen-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 3-hexan-1-ol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'