missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Aminocyclohexanecarboxylic acid, 97%

Product Code. p-7030038
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11406460

Brand: Thermo Scientific Alfa Aesar H27567.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3685-23-2 | C7H13NO2 | 143.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3685-23-2
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD00191730
InChI Key DRNGLYHKYPNTEA-UHFFFAOYSA-N
Synonym 4-aminocyclohexanecarboxylic acid, trans-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, trans-4-amino-cyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-aminocyclohexane carboxylic acid, 1s,4s-4-aminocyclohexanecarboxylic acid
PubChem CID 171560
IUPAC Name 4-aminocyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)N

Specifications

Melting Point 299°C to 301°C
Quantity 1 g
Formula Weight 143.19
Percent Purity 97%
Chemical Name or Material cis-4-Aminocyclohexanecarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.