missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Octene, 97%, Thermo Scientific Chemicals

Product Code. 11374838
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

cis-4-octene is used as a chain transfer agent in polymerization reactions, and as a depolymerizing agent in epoxy-functionalized oligomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7642-15-1
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00067484
InChI Key IRUCBBFNLDIMIK-FPLPWBNLSA-N
Synonym cis-4-octene, z-oct-4-ene, z-4-octene, 4-octene, z, unii-19430whf5e, 4z-4-octene, 4z-oct-4-ene, 4-octene, 4z, cis-4-octene 1g, unii-68k818w2zc component
PubChem CID 5364446
SMILES CCC\C=C/CCC

Specifications

Quantity 2 g
UN Number UN3295
Solubility Information Immiscible with water.
IUPAC Name (Z)-oct-4-ene
Formula Weight 112.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material cis-4-Octene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.