missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ cis-6-Hydroxynorketamine hydrochloride
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
cis-6-Hydroxynorketamine hydrochloride is a metabolite of ketamine . Increases AMPA receptor-mediated excitatory post-synaptic potentials in the CA1 region of hippocampal slices. Exhibits antidepressive effects in vivo. 2S,6S-enantiomer and 2R,6R-enantiomer also available.
Specifications
Specifications
| Chemical Name or Material | cis-2-Amino-2-(2-chlorophenyl)-6-hydroxycyclohexanone hydrochloride |
| CAS | 2319601-04-0 |
| Quantity | 10 mg |
| Target | Additional NMDA Receptor Compounds |
| Molecular Formula | C12H14ClNO2.HCl |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction