missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis,cis-1,5-Cyclooctadiene, 99%, stab. with 50-200ppm Irganox 1076, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1000 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | °C to 70°C |
Density | 0.880 |
Boiling Point | 151°C to 152°C |
Flash Point | 32°C (89°F) |
Assay Percent Range | 0.99 |
Refractive Index | 1.494 |
Quantity | 250 mL |
UN Number | UN2520 |
Beilstein | 1209288 |
Sensitivity | Air Sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'