missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-Diamminedichloroplatinum(II), Pt 64.5% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Being a potent anticancer agent that blocks DNA synthesis, the pharmaceutical compositions are used in chemotherapy. First member of platinum-containing anti-cancer drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 270°C (decomposition) |
Color | Yellow |
Physical Form | Micro-crystals |
Quantity | 5 g |
Solubility Information | Soluble in DMF. Insoluble in most common solvents |
Formula Weight | 300.06 |
Odor | Odorless |
Appearance | Yellow micro crystals |
Chemical Name or Material | cis-Diamminedichloroplatinum(II), Potent anticancer agent that blocks DNA synthesis |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'