Learn More
cis-Diamminedichloroplatinum(II), Pt 64.5% min, Thermo Scientific Chemicals
Potent Anticancer Agent That Blocks DNA Synthesis.
£389.00 - £1041.00
Chemical Identifiers
CAS | 15663-27-1 |
---|---|
Molecular Formula | Cl2H6N2Pt |
Molecular Weight (g/mol) | 300.04 |
MDL Number | MFCD00011623 |
InChI Key | BSJGASKRWFKGMV-UHFFFAOYSA-L |
Synonym | Cisplatin |
IUPAC Name | dichloroplatinumbis(ylium) diamine |
SMILES | N.N.Cl[Pt++]Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11335978
|
Thermo Scientific Alfa Aesar
010471.03 |
1 g |
£389.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11325978
|
Thermo Scientific Alfa Aesar
010471.06 |
5 g |
£1041.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Being a potent anticancer agent that blocks DNA synthesis, the pharmaceutical compositions are used in chemotherapy. First member of platinum-containing anti-cancer drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15663-27-1 | |
300.04 | |
BSJGASKRWFKGMV-UHFFFAOYSA-L | |
dichloroplatinumbis(ylium) diamine |
Cl2H6N2Pt | |
MFCD00011623 | |
Cisplatin | |
N.N.Cl[Pt++]Cl |
Specifications
270°C (decomposition) | |
Yellow | |
Cl2H6N2Pt | |
Cisplatin | |
BSJGASKRWFKGMV-UHFFFAOYSA-L | |
dichloroplatinumbis(ylium) diamine | |
300.06 | |
Yellow micro crystals |
15663-27-1 | |
Micro-crystals | |
MFCD00011623 | |
Soluble in DMF. Insoluble in most common solvents | |
N.N.Cl[Pt++]Cl | |
300.04 | |
Odorless | |
cis-Diamminedichloroplatinum(II), Potent anticancer agent that blocks DNA synthesis |
Safety and Handling
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P362-P501c
H300-H312+H332-H315-H318-H335-H350
missing translation for 'dotInformation' : DOT Class: 6.1, Packing Group: II
missing translation for 'einecsNumber' : 239-733-8
missing translation for 'rtecsNumber' : TP2450000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only