Learn More
cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis), Thermo Scientific Chemicals
£79.90 - £722.00
Chemical Identifiers
CAS | 15663-27-1 |
---|---|
Molecular Formula | Cl2H6N2Pt |
Molecular Weight (g/mol) | 300.04 |
MDL Number | MFCD00011623 |
InChI Key | BSJGASKRWFKGMV-UHFFFAOYSA-L |
Synonym | cis-DDP |
IUPAC Name | dichloroplatinumbis(ylium) diamine |
SMILES | N.N.Cl[Pt++]Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10246680
|
Thermo Scientific Acros
193762500 |
250 mg | Glass bottle |
£79.90
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10791155
|
Thermo Scientific
193760010 |
1 g | Glass bottle |
£222.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10371614
|
Thermo Scientific Acros
193760050 |
5 g | Glass bottle |
£722.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
15663-27-1 | |
300.04 | |
BSJGASKRWFKGMV-UHFFFAOYSA-L | |
dichloroplatinumbis(ylium) diamine |
Cl2H6N2Pt | |
MFCD00011623 | |
cis-DDP | |
N.N.Cl[Pt++]Cl |
Specifications
277.0°C | |
White | |
64.0 to 66% (Pt) | |
Pt(NH3)2Cl2 | |
15, 2316 | |
Solubility in water: 0.25g/100mL (25°C) | |
N.N.Cl[Pt++]Cl | |
300.04 | |
99.99% | |
Authentic |
15663-27-1 | |
Powder | |
Cl2H6N2Pt | |
MFCD00011623 | |
cis-DDP | |
BSJGASKRWFKGMV-UHFFFAOYSA-L | |
dichloroplatinumbis(ylium) diamine | |
300.05 | |
Trace Metal Basis | |
cis-Dichlorodiamineplatinum(II), 99.99% |
Safety and Handling
GHS H Statement
May cause cancer.
Fatal if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 239-733-8
missing translation for 'rtecsNumber' : TP2450000
missing translation for 'tsca' : TSCA
- cis-Dichlorodiamineplatinum(II)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H350-May cause cancer.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H300-Fatal if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only