missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cisplatin, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Potent anticancer agent that blocks DNA synthesis. Induces apoptosis via p53-dependent and -independent mechanisms. Inhibits X-linked inhibitor of apoptosis protein (XIAP) expression and activates caspase-3. In certain glioma cell lines, sensitizes cells to TNF-α-induced apoptosis.
Specifications
Specifications
CAS | 15663-27-1 |
Quantity | 50 mg |
Molecular Formula | Cl2H6N2Pt |
MDL Number | MFCD00011623 |
Synonym | cis-Diaminodichloroplatinum |
InChI Key | BSJGASKRWFKGMV-UHFFFAOYSA-L |
SMILES | N.N.Cl[Pt++]Cl |
IUPAC Name | dichloroplatinumbis(ylium) diamine |
Molecular Weight (g/mol) | 300.04 |
Formula Weight | 300.05 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction