missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Citadel 2000 Tissue Processor
The Epredia™ Citadel 2000 Tissue Processor is a low cost, reliable, time-tested carousel-type tissue processor with convenient touch key operation and a 110-cassette capacity.
£16447.50
Specifications
Product Type | Tissue Processor |
---|---|
Basket Capacity | 110 (Organised); 70 (Random) |
Reagent Capacity | 2 L |
Frequency | 50/60 Hz |
Product Code | Brand | Voltage | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Voltage | Price | Quantity & Availability | |||||
12612613
|
Epredia™
69800004 |
220/240 VAC | N/A | ||||||
Description
The compact design of the Citadel 2000 carousel tissue processor, coupled with its proven processing quality, ease of operation and excellent chemical resistance explains why it has become the tried and trusted choice for thousands of laboratories worldwide.
Features:
- Specimens are automatically raised from their reagent containers every 10 minutes and then reimmersed
- Optional vacuum unit for efficient and rapid wax infiltration
- Unique touch key pad incorporates all processing functions
- Up to four complete schedules can be programmed
- Programmable delayed start time
- Loading and unloading of specimens is made easy with the specially designed tissue basket hanger
- Random baskets holding 70 cassettes or 110 cassette organiser baskets
- Smooth surfaces and contoured design facilitate easy cleaning
- Integral fume hood greatly reduces release of vapours
- Easy change of reagent containers
Physical Specifications:
- Electrical Requirements: 100-240 V, 50/60 Hz
- Diameter: 68 cm (27 in.)
- Height: 62 cm (25 in.)
- Weight: 106 lb. (48 kg)
Includes:
- Basic instrument
- Instruction manual
- Ten reagent vessels and two paraffin baths
Specifications
Tissue Processor | |
110 (Organised); 70 (Random) | |
2 L | |
50/60 Hz |