missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citraconic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Citraconic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Citraconic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
Citraconic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 88°C to 91°C |
Density | 1.62 |
Quantity | 10 g |
Beilstein | 1722679 |
Sensitivity | Hygroscopic |
Merck Index | 14,2321 |
Solubility Information | Soluble in water. |
Formula Weight | 130.1 |
Percent Purity | ≥98% |
Chemical Name or Material | Citraconic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'