missing translation for 'onlineSavingsMsg'
Learn More
Citraconic acid, 99+%, Thermo Scientific Chemicals Product Code.: 10530181

Citraconic acid, 99+%, Thermo Scientific Chemicals

Product Code. 10530181 Shop All Thermo Scientific Products
100 g, Plastic bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10530181

Brand: Thermo Scientific Acros 110431000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-23-7
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.10
MDL Number MFCD00078086
InChI Key HNEGQIOMVPPMNR-IHWYPQMZSA-N
Synonym 2-methylmaleate, citraconate 2-, z-2-methylbut-2-enedioate
PubChem CID 5461090
ChEBI CHEBI:30719
IUPAC Name (Z)-2-methylbut-2-enedioate
SMILES C\C(=C\C(O)=O)C(O)=O

Specifications

Melting Point 88.0°C to 92.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula CH3C(CO2H)=CHCO2H
Quantity 100 g
Beilstein 02,768
Merck Index 15,232
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol and ether,slightly soluble in chloroform,insoluble in benzene and petroleum ether
Formula Weight 130.1
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Citraconic acid, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466401 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog Number110431000,110430250,110430050,11043023
Lot NumberA0429907 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog Number110431000,110430250,110430050,11043023
Lot NumberA0454702 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number110431000,110430250,110430050,11043023
Lot NumberA0397116 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number110431000,110430250,110430050,11043023
Lot NumberA0401753 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number110431000,110430250,110430050,11043023
compliance-icons
Product Identifier
  • Citraconic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Citraconic acid, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.