Learn More
Citraconic anhydride, 98%, Thermo Scientific Chemicals
£20.50 - £164.82
Chemical Identifiers
CAS | 616-02-4 |
---|---|
Molecular Formula | C5H4O3 |
Molecular Weight (g/mol) | 112.084 |
MDL Number | MFCD00005522 |
InChI Key | AYKYXWQEBUNJCN-UHFFFAOYSA-N |
Synonym | citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx |
PubChem CID | 12012 |
IUPAC Name | 3-methylfuran-2,5-dione |
SMILES | CC1=CC(=O)OC1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11341857
|
Thermo Scientific Alfa Aesar
L05238.14 |
25 g |
£20.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357355
|
Thermo Scientific Alfa Aesar
L05238.22 |
100 g |
£46.83
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367355
|
Thermo Scientific Alfa Aesar
L05238.36 |
500 g |
£164.82
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Citraconic anhydride is used as a monomer for specialty unsaturated polyester resins. It is used to prepare bicyclic pyrrolidines, maleimides and co- and terpolymers. It is also used for the protection of N-terminal amino acids. Further, it is involved in the preparation of clothing ping acid and its amide. In addition to this, it is selectively used for the isolation of histidyl peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCitraconic anhydride is used as a monomer for specialty unsaturated polyester resins. It is used to prepare bicyclic pyrrolidines, maleimides and co- and terpolymers. It is also used for the protection of N-terminal amino acids. Further, it is involved in the preparation of clothing ping acid and its amide. In addition to this, it is selectively used for the isolation of histidyl peptides.
Solubility
Miscible with water, alcohol, acetone and ether.
Notes
Moisture sensitive. Incompatible with bases, oxidizing agents and reducing agents.
Chemical Identifiers
616-02-4 | |
112.084 | |
AYKYXWQEBUNJCN-UHFFFAOYSA-N | |
12012 | |
CC1=CC(=O)OC1=O |
C5H4O3 | |
MFCD00005522 | |
citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx | |
3-methylfuran-2,5-dione |
Specifications
616-02-4 | |
1.243 | |
101°C (213°F) | |
1.471 | |
UN2810 | |
Moisture sensitive | |
Miscible with water,alcohol,acetone and ether. | |
CC1=CC(=O)OC1=O | |
112.084 | |
112.08 | |
Citraconic anhydride |
7°C to 8°C | |
213°C to 214°C | |
C5H4O3 | |
MFCD00005522 | |
1835 | |
citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx | |
AYKYXWQEBUNJCN-UHFFFAOYSA-N | |
3-methylfuran-2,5-dione | |
12012 | |
98% |
Safety and Handling
GHS H Statement
H311-H315-H319
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
H311-H314-H317-H334
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 210-459-0
missing translation for 'rtecsNumber' : GE6825000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only