Learn More
Citrate Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand:  Fisher Chemical A 015 500
 View more versions of this product
Product Code. 12942115
  • £104.00 / 500mL
Explore more special offers
Specifications
Citrate
Bottle
Single-component Standard Solution
H2O
1g/L
500 mL
C6H5O73-