missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, 99.5 to 100.5% (based on Anhydrous substance), Honeywell Fluka™
BIOPHARMA

Product Code. 15682880 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
100g
1kg
2.5kg
25kg
Pack of 6
This item is not returnable. View return policy

Product Code. 15682880

Brand: Honeywell-Fluka 271092.5KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Meets analytical specification of BP, Ph. Eur., USP, E330, Anhydrous, 99.5-100.5% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Linear Formula HOC(COOH)(CH2COOH)2
UN Number NONH for all modes of transport
Beilstein 782061
Solubility Information Soluble in water, 383g/L at 25°C, Sparingly soluble in Diethyl Ether, Freely soluble in alcohol
Formula Weight 192.12g/mol
Percent Purity 99.5 to 100.5% (based on anhydrous substance)
Quantity 2.5 kg
Explosion Limit 8% (65°F)
Physical Form Anhydrous
Residue after Ignition ≤0.05% (as SO4)
Chemical Name or Material Citric Acid, Meets analytical specification of Ph. Eur.BP, USP
Show More Show Less
compliance-icons
Product Identifier
  • Citric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Citric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.