missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, 99.5 to 100.5% (based on Anhydrous substance), Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Linear Formula | HOC(COOH)(CH2COOH)2 |
UN Number | NONH for all modes of transport |
Beilstein | 782061 |
Solubility Information | Soluble in water, 383g/L at 25°C, Sparingly soluble in Diethyl Ether, Freely soluble in alcohol |
Formula Weight | 192.12g/mol |
Percent Purity | 99.5 to 100.5% (based on anhydrous substance) |
Quantity | 25 kg |
Explosion Limit | 8% (65°F) |
Physical Form | Anhydrous |
Residue after Ignition | ≤0.05% (as SO4) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'