missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, 99+%, Thermo Scientific Chemicals

£15.68 - £103.52

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11498573
View Documents
Thermo Scientific Alfa Aesar
A10395.30
250 g
£15.68
250g
Please to purchase this item. Need a web account? Register with us today!
11488573
View Documents
Thermo Scientific Alfa Aesar
A10395.0B
1000 g
£34.27
1000g
Please to purchase this item. Need a web account? Register with us today!
11408583
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A10395.0I
5000 g
£103.52
5000g
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

Citric acid is the most widely used organic acid and pH-control agent in foods, beverages, pharmaceuticals and technical applications. Used in pharmaceutical preparations, especially in effervescent tablets. It is used as a natural preservative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citric acid is the most widely used organic acid and pH-control agent in foods, beverages, pharmaceuticals and technical applications. Used in pharmaceutical preparations, especially in effervescent tablets. It is used as a natural preservative.

Solubility
Soluble in water, alcohol, ether, dimethylsufoxide and ethylacetate.

Notes
Hygroscopic. Incompatible with alkalies, bases, metal nitrates and oxidizing agents.
Especificaciones

Identificadores de productos químicos

77-92-9
192.12
KRKNYBCHXYNGOX-UHFFFAOYSA-N
311
2-hydroxypropane-1,2,3-tricarboxylic acid
C6H8O7
MFCD00011669
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
CHEBI:30769
OC(=O)CC(O)(CC(O)=O)C(O)=O

Especificaciones

1.6
153°C to 154°C
MFCD00011669
14,2326
Soluble in water,alcohol,ether,dimethylsufoxide and ethylacetate.
OC(=O)CC(O)(CC(O)=O)C(O)=O
192.12
CHEBI:30769
≥99%
Odorless
77-92-9
C6H8O7
782061
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
KRKNYBCHXYNGOX-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid
311
192.13
1.665
Citric acid
Videos
Seguridad y manipulación

Seguridad y manipulación

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H335-H500

missing translation for 'einecsNumber' : 201-069-1

missing translation for 'rtecsNumber' : GE7350000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Palabra de advertencia
  • Warning

Clasificación
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Otros peligros
  • MIXTURE LIST-Contains : Citric acid

SDS
Documentos

Documentos

Certificados

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado