missing translation for 'onlineSavingsMsg'
Learn More
Citric Acid, Anhydrous, 99%, Pure, Thermo Scientific Chemicals Product Code.: 10338903

Citric Acid, Anhydrous, 99%, Pure, Thermo Scientific Chemicals

Product Code. 10338903
5 kg, Plastic Bottle
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10338903

Brand: Thermo Scientific Acros 110450050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxyShow MoreShow Less
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Melting Point 153.0°C
Color White
Assay Percent Range 99%
Linear Formula HO2CCH2C(OH)(CO2H)CH2CO2H
Beilstein 03, 556
Merck Index 15, 2325
Solubility Information Solubility in water: 750g/L (20°C). Other solubilities: soluble in alcohol and ether, badly soluble in diethyl ether, insoluble in benzene and chloroform
Formula Weight 192.13
Percent Purity 99%
Grade Pure
Quantity 5 kg
Flash Point 100°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Physical Form Crystalline
Chemical Name or Material Citric acid, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0427603 Certificate TypeCertificate of Analysis Date28/08/2023 Catalog Number110450250,110450050,110450010
Lot NumberA0453307 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number110450250,110450050,110450010
Lot NumberA0393282 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number110450250,110450050,110450010
Lot NumberA0393283 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number110450250,110450050,110450010
Lot NumberA0410335 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number110450250,110450050,110450010
compliance-icons
Product Identifier
  • Citric acid anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Citric Acid, Anhydrous, 99%, Pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.