missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, diammonium salt, 98%, pure

Product Code. 10217713 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10217713

Brand: Thermo Scientific Acros 392505000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3012-65-5 | C6H14N2O7 | 226.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3012-65-5
Molecular Formula C6H14N2O7
Molecular Weight (g/mol) 226.19
MDL Number MFCD00013068
InChI Key YXVFQADLFFNVDS-UHFFFAOYSA-N
Synonym ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss
PubChem CID 13710713
ChEBI CHEBI:63076
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES [NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O

Specifications

Color White
Density 1.5000g/mL
Infrared Spectrum Authentic
Assay Percent Range 0.98
Packaging Plastic bottle
Quantity 500 g
Merck Index 15, 509
Specific Gravity 1.5
Solubility Information Solubility in water: soluble.
Formula Weight 226.19
Percent Purity 0.98
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material Citric acid, diammonium salt
Show More Show Less
compliance-icons
Product Identifier
  • Citric acid, diammonium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.