missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric acid diammonium salt, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
Citric acid diammonium salt is used in pharmaceuticals, rust proofing, cotton printing and plasticizers. It is used as a reagent for the determination of phosphate in fertilizers, and as a buffering agent, chelating agent and source of nitrogen in chicken food supplements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Density | 1.48 g/mL |
Odor | Odorless |
Linear Formula | HOC(CO2H)(CH2CO2NH4)2 |
Quantity | 500 g |
Merck Index | 14512 |
Solubility Information | Soluble in water. Slightiy soluble in alcohol. |
Formula Weight | 226.19 |
Percent Purity | ≥99% |
Physical Form | Granules |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'