Learn More
Citric acid diammonium salt, ACS, 98.0-103.0%, Thermo Scientific Chemicals
£25.00 - £136.00
Chemical Identifiers
CAS | 3012-65-5 |
---|---|
Molecular Formula | C6H14N2O7 |
Molecular Weight (g/mol) | 226.19 |
MDL Number | MFCD00013068 |
InChI Key | YXVFQADLFFNVDS-UHFFFAOYSA-N |
Synonym | ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss |
PubChem CID | 13710713 |
ChEBI | CHEBI:63076 |
IUPAC Name | azane;2-hydroxypropane-1,2,3-tricarboxylic acid |
SMILES | [NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11343268
|
Thermo Scientific Alfa Aesar
036219.22 |
100 g |
£25.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11353268
|
Thermo Scientific Alfa Aesar
036219.36 |
500 g |
£53.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11333268
|
Thermo Scientific Alfa Aesar
036219.A3 |
2 kg |
£136.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Citric acid diammonium salt is used in pharmaceuticals, rust proofing, cotton printing and plasticizers. It is also used as a reagent for the determination of phosphate in fertilizers, and as a buffering agent, chelating agent and source of nitrogen in chicken food supplements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3012-65-5 | |
226.19 | |
YXVFQADLFFNVDS-UHFFFAOYSA-N | |
13710713 | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid |
C6H14N2O7 | |
MFCD00013068 | |
ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss | |
CHEBI:63076 | |
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O |
Specifications
3012-65-5 | |
Odorless | |
MFCD00013068 | |
ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss | |
YXVFQADLFFNVDS-UHFFFAOYSA-N | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid | |
13710713 | |
226.19 | |
ACS | |
Citric acid diammonium salt |
1.48 g/mL | |
C6H14N2O7 | |
14512 | |
Soluble in water. Slightiy soluble in alcohol. | |
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O | |
226.19 | |
CHEBI:63076 | |
98.0 to 103.0% | |
Granules |
Safety and Handling
GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335
missing translation for 'einecsNumber' : 221-146-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only