Learn More
Citric acid, Electrophoresis Grade, 99.5+%, Thermo Scientific Chemicals
£16.98 - £82.91
Chemical Identifiers
CAS | 77-92-9 |
---|---|
Molecular Formula | C6H8O7 |
Molecular Weight (g/mol) | 192.12 |
MDL Number | MFCD00011669 |
InChI Key | KRKNYBCHXYNGOX-UHFFFAOYSA-N |
Synonym | citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy |
PubChem CID | 311 |
ChEBI | CHEBI:30769 |
IUPAC Name | 2-hydroxypropane-1,2,3-tricarboxylic acid |
SMILES | OC(=O)CC(O)(CC(O)=O)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15466319
|
Thermo Scientific Alfa Aesar
J64322.22 |
100 g |
£16.98
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15476319
|
Thermo Scientific Alfa Aesar
J64322.36 |
500 g |
£33.37
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15486319
|
Thermo Scientific Alfa Aesar
J64322.A3 |
2 kg |
£82.91
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Citric acid is a starting point in the Krebs cycle and is a key intermediate in metabolism. It has anticoagulant activity; as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot. It may be used to adjust pH and as a sequestering agent for the removal of trace metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCitric acid is a starting point in the Krebs cycle and is a key intermediate in metabolism. It has anticoagulant activity; as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot. It may be used to adjust pH and as a sequestering agent for the removal of trace metals.
Solubility
Soluble in water. Also soluble in alcohol and methanol. Insoluble in chloroform and benzene.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Chemical Identifiers
77-92-9 | |
192.12 | |
KRKNYBCHXYNGOX-UHFFFAOYSA-N | |
311 | |
2-hydroxypropane-1,2,3-tricarboxylic acid |
C6H8O7 | |
MFCD00011669 | |
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy | |
CHEBI:30769 | |
OC(=O)CC(O)(CC(O)=O)C(O)=O |
Specifications
1.6 | |
153°C to 154°C | |
C6H8O7 | |
782061 | |
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy | |
KRKNYBCHXYNGOX-UHFFFAOYSA-N | |
2-hydroxypropane-1,2,3-tricarboxylic acid | |
311 | |
192.13 | |
Electrophoresis | |
Odorless | |
Hygroscopic |
77-92-9 | |
White | |
MFCD00011669 | |
14,2326 | |
Soluble in water. Also soluble in alcohol and methanol. Insoluble in chloroform and benzene. | |
OC(=O)CC(O)(CC(O)=O)C(O)=O | |
192.12 | |
CHEBI:30769 | |
≥99.5% | |
1.665 | |
Powder | |
Citric acid |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335-H500
missing translation for 'einecsNumber' : 201-069-1
missing translation for 'rtecsNumber' : GE7350000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.