missing translation for 'onlineSavingsMsg'
Learn More
Citric acid, for molecular biology, anhydrous, Rnase and Protease free Product Code.: 10565871

Citric acid, for molecular biology, anhydrous, Rnase and Protease free

Product Code. 10565871
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy

Product Code. 10565871

Brand: Thermo Scientific Acros 329860010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77-92-9 | C6H8O7 | 192.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for molecular biology
TRUSTED_SUSTAINABILITY

Specifications

CAS 77-92-9
Melting Point 153.0°C
Color White
Molecular Formula C6H8O7
Linear Formula HO2CCH2C(OH)(CO2H)CH2CO2H
MDL Number MFCD00011669
Beilstein 03, 556
Merck Index 15, 2325
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
Solubility Information Solubility in water: 750g/L (20°C). Other solubilities: soluble in alcohol and ether, badly soluble in diethyl ether, insoluble in benzene and chloroform
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Weight (g/mol) 192.12
PubChem CID 311
ChEBI CHEBI:30769
Formula Weight 192.13
Percent Purity ≥99%
Grade Molecular Biology
Quantity 1 kg
Flash Point 100°C
Infrared Spectrum Authentic
Physical Form Crystalline Solid
Chemical Name or Material Citric acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464381 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog Number329860010
Lot NumberA0433187 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number329860010
Lot NumberA0393432 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number329860010
Lot NumberA0414477 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number329860010
Lot NumberA0429727 Certificate TypeCertificate of Analysis Date04/05/2022 Catalog Number329860010
compliance-icons
Product Identifier
  • Citric acid anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Citric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Citric acid, for molecular biology, anhydrous, Rnase and Protease free >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.