missing translation for 'onlineSavingsMsg'
Learn More
Citric acid monohydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals Product Code.: 11348098

Citric acid monohydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals

Product Code. 11348098
2 kg
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11348098

Brand: Thermo Scientific Alfa Aesar 036665.A3

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Citric acid monohydrate is used in the preparation of citrate buffer in platelets for intravital microscopy. It acts as a pH-control agent in foods, beverages and pharmaceuticals applications. It acts as an iron chelator. In animals, it improves the utilization of nutritional calcium. It is a useful buffer component for antigen and epitope unmasking. It is also used as an acidifier, a flavoring agent and a chelating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.138
MDL Number MFCD00149972
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate uspShow MoreShow Less
PubChem CID 22230
ChEBI CHEBI:31404
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O

Specifications

Color White
Merck Index 14,2326
Solubility Information Soluble in water,ethanol and ether. Partially soluble in methanol,diethyl ether. Insoluble in n-octanol.
Formula Weight 210.14 (192.13 Anhydrous)
Percent Purity 99.0 to 102.0%
Grade ACS
Density 1.542 g/mL
Quantity 2 kg
Odor Odorless
Physical Form Solid
Sensitivity Hygroscopic
Chemical Name or Material Citric acid monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY03C001 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog Number036665.22,036665.A3,036665.36
Lot NumberS17K024 Certificate TypeCertificate of Analysis Date27/06/2024 Catalog Number036665.22,036665.A3,036665.36
Lot NumberP27K006 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number036665.22,036665.A3,036665.36
Lot NumberM22K005 Certificate TypeCertificate of Analysis Date26/01/2024 Catalog Number036665.22,036665.A3,036665.36
Lot NumberW20J038 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog Number036665.22,036665.A3,036665.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Citric acid monohydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.