missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric acid monohydrate, buffer substance, 99.5-102%, Honeywell Fluka™
Click to view available options
25 kg
Description
Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 5949-29-1 |
Molecular Formula | C6H10O8 |
InChI Key | YASYEJJMZJALEJ-UHFFFAOYSA-N |
SMILES | O.OC(=O)CC(O)(CC(O)=O)C(O)=O |
IUPAC Name | 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate |
Molecular Weight (g/mol) | 210.14 |
Quantity | 25 kg |
Chemical Name or Material | Citric acid monohydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'