missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid Monohydrate, Extra Pure, SLR, Granular, Fisher Chemical™
39.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Specifications
pH | 1.8 |
Melting Point | 135°C |
Color | White |
Formula Weight | 210.15g/mol |
Quantity | 500 g |
Packaging | Plastic powder jar |
Physical Form | Solid |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Citric acid monohydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Citric acid monohydrate, Citric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'