missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid Monohydrate, Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
71.51 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 kg
25 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Linear Formula | HOC(COOH)(CH2COOH)2 · H2O |
UN Number | NONH for all modes of transport |
Beilstein | 4018641 |
Solubility Information | Soluble in ethyl acetate (5.28g/100g), Amyl Acetate(5.98g/100g), diethyl ether(2.17g/100g), methanol(197g/100g), chloroform(0.007g/100 g) |
Formula Weight | 210.14g/mol |
Quantity | 1 kg |
Flash Point | 173°C (343.4°F) |
Chemical Name or Material | Citric Acid Monohydrate, Tested according to Ph.Eur. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'