missing translation for 'onlineSavingsMsg'
Learn More
Citric acid monohydrate, for HPLC, Thermo Scientific Chemicals Product Code.: 16514274
compliance-icons

Citric acid monohydrate, for HPLC, Thermo Scientific Chemicals

Product Code. 16514274
100 g
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy

Product Code. 16514274

Brand: Thermo Scientific Acros 464541000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5949-29-1
Melting Point 135.0°C to 152.0°C
Molecular Formula C6H10O8
Linear Formula HO2CCH2C(OH)(CO2H)CH2CO2H·H2O
Solubility Information Solubility aq. soln.: 676 g/L (25°C). Other solubilities: 2.17 g/100 g ether - 0.007 g/100 g chloroform, 15.43 g/100 g amyl alcohol - 5.98 g/100 g amyl acetate, 5.28 g/100 g ethyl acetate - 197 g/100 g methanol, 62.8 g/100 g propanol
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
SMILES O.OC(=O)CC(O)(CC(O)=O)C(O)=O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
Molecular Weight (g/mol) 210.14
Formula Weight 210.15
Grade HPLC
Quantity 100 g
Flash Point 174°C
Chemical Name or Material Citric acid monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442881 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number464541000
Lot NumberA0459487 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog Number464541000
Lot NumberA0453034 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number464541000
Lot NumberA0449911 Certificate TypeCertificate of Analysis Date30/01/2023 Catalog Number464541000
Lot NumberA0410720 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number464541000
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Citric acid monohydrate, for HPLC, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.