missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, Powder, U.S.P., J.T. Baker™
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Bottle
Specifications
Specifications
Formula Weight | 192.12 |
Grade | USP |
Quantity | 2 kg |
Packaging | Poly Bottle |
Physical Form | Powder |
Specific Gravity | 1L = 1.44kg |
Chemical Name or Material | Citric Acid |
Safety and Handling
Product Identifier
- Citric acid, anhydrous
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Citric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'