missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, SPEX CertiPrep™
Click to view available options
Quantity:
1 mL
Packaging:
2 mL amber prescored glass ampule
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Citric Acid | 77-92-9 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
CAS | 77-92-9 |
Molecular Formula | C6H8O7 |
MDL Number | MFCD00011669 |
Synonym | citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy |
InChI Key | KRKNYBCHXYNGOX-UHFFFAOYSA-N |
SMILES | OC(=O)CC(O)(CC(O)=O)C(O)=O |
IUPAC Name | 2-hydroxypropane-1,2,3-tricarboxylic acid |
Molecular Weight (g/mol) | 192.12 |
Concentration or Composition (by Analyte or Components) | 1000 μg/mL |
Quantity | 1 mL |
Show More |
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'