missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, triammonium salt, 97+%, pure

Product Code. p-3593406 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 10320624

Brand: Thermo Scientific Acros 391410500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3458-72-8 | C6H20N3O7+3 | 246.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3458-72-8
Molecular Formula C6H20N3O7+3
Molecular Weight (g/mol) 246.24
MDL Number MFCD00036406
InChI Key YWYZEGXAUVWDED-UHFFFAOYSA-Q
Synonym ammonium citrate
PubChem CID 131675891
IUPAC Name azane;[3-carboxy-1,3-dihydroxy-4-[oxonio(oxoniumylidene)methyl]butylidene]oxidanium
SMILES C(C(=[OH+])[OH2+])C(CC(=[OH+])O)(C(=O)O)O.N.N.N

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Plastic bottle
Quantity 50 g
Formula Weight 243.22
Percent Purity ≥97.0%
Grade Extra Pure
Physical Form Crystals
Chemical Name or Material Citric acid, triammonium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.