missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric acid, trisodium salt dihydrate, 99%, pure, Thermo Scientific Chemicals
£15.40 - £487.43
Chemical Identifiers
CAS | 1545801 |
---|---|
Molecular Formula | C6H9Na3O9 |
Molecular Weight (g/mol) | 294.1 |
MDL Number | MFCD00150031 |
InChI Key | NLJMYIDDQXHKNR-UHFFFAOYSA-K |
Synonym | trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate |
PubChem CID | 71474 |
ChEBI | CHEBI:32142 |
IUPAC Name | trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate |
SMILES | C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10083010
|
Thermo Scientific Acros
227130050 |
5 g | Glass Bottle | N/A | |||||
10338460
|
Thermo Scientific Acros
227135000 |
500 g | Plastic Bottle |
£25.93
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10134520
|
Thermo Scientific Acros
227130010 |
1 kg | Plastic Bottle |
£39.50
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10083163
|
Thermo Scientific Acros
227130051 |
5 kg | Plastic bottle |
£127.64
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13438488
|
Thermo Scientific Acros
227130250 |
25 kg | Fibre drum |
£487.43
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1545801 | |
294.1 | |
NLJMYIDDQXHKNR-UHFFFAOYSA-K | |
71474 | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate |
C6H9Na3O9 | |
MFCD00150031 | |
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate | |
CHEBI:32142 | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] |
Specifications
1545801 | |
White | |
C6H9Na3O9 | |
MFCD00150031 | |
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate | |
NLJMYIDDQXHKNR-UHFFFAOYSA-K | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate | |
71474 | |
294.09 | |
Pure | |
Citric acid, trisodium salt dihydrate |
>300.0°C | |
Authentic | |
NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O | |
15, 8737 | |
Solubility in water: 720g/L (25°C). Other solubilities: insoluble in alcohol | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] | |
294.1 | |
CHEBI:32142 | |
0.99 | |
Crystalline Powder or Crystals |
Safety and Handling
Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated.
NFPA
Health:
Signal Word: CAUTION!
Product Identifier
- Citric acid, trisodium salt dihydrate
RUO – Research Use Only