missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric acid, trisodium salt dihydrate, ACS reagent, Thermo Scientific Chemicals
15.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | >300.0°C |
Packaging | Plastic Bottle |
Linear Formula | NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O |
Quantity | 250 g |
Merck Index | 15,8737 |
Solubility Information | Solubility in water: 720g/L (25°C). Other solubilities: insoluble in alcohol |
Formula Weight | 294.09 |
Grade | ACS Reagent |
Insoluble Matter | 0.005% max |
Chemical Name or Material | Citric Acid, Trisodium Salt Dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'