missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, U.S.P. - F.C.C., J.T. Baker™
£176.00 - £436.00
Chemical Identifiers
CAS | 5949-29-1 |
---|---|
Molecular Formula | C6H10O8 |
Molecular Weight (g/mol) | 210.138 |
InChI Key | YASYEJJMZJALEJ-UHFFFAOYSA-N |
Synonym | citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp |
PubChem CID | 22230 |
ChEBI | CHEBI:31404 |
IUPAC Name | 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate |
SMILES | C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15567774
|
Avantor J.T.Baker
0119-01 |
500 g | Wide Mouth Amber Glass Bottle |
£176.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15577774
|
Avantor J.T.Baker
0119-05 |
2.5 kg | Poly Bottle |
£436.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
5949-29-1 | |
210.138 | |
citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp | |
CHEBI:31404 | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O |
C6H10O8 | |
YASYEJJMZJALEJ-UHFFFAOYSA-N | |
22230 | |
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate |
Specifications
5949-29-1 | |
citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O | |
210.138 | |
CHEBI:31404 | |
FCC/USP | |
Citric Acid |
C6H10O8 | |
YASYEJJMZJALEJ-UHFFFAOYSA-N | |
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate | |
22230 | |
210.14 | |
1L = 1.54kg |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Citric Acid Monohydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Citric acid, monohydrate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title