missing translation for 'onlineSavingsMsg'
Learn More
Citrinin, 98%, Thermo Scientific Chemicals Product Code.: 10105190

Citrinin, 98%, Thermo Scientific Chemicals

Product Code. 10105190
5mg; Glass bottle packed in Plastic container
Click to view available options
5mg; Glass bottle packed in Plastic container
This item is not returnable. View return policy

Product Code. 10105190

Brand: Thermo Scientific Acros 229580050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 518-75-2
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C13H14O5
MDL Number MFCD00006912
Quantity 5 mg
Merck Index 15, 2326
Specific Rotation − 30.30
Synonym citrinin, antimycin, --citrinin, citriain, 3r,4s-4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid, unii-3s697x6snz, ccris 175, 3h-2-benzopyran-7-carboxylic acid, 4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-, 3r-trans, 3r-trans-4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid, 4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid
InChI Key CBGDIJWINPWWJW-IYSWYEEDSA-N
SMILES CC1C(OC=C2C1=C(C(=C(C2=O)C(=O)O)O)C)C
IUPAC Name (3R,4S)-6-hydroxy-3,4,5-trimethyl-8-oxo-3,4-dihydroisochromene-7-carboxylic acid
Molecular Weight (g/mol) 250.25
PubChem CID 54680783
Formula Weight 250.25
Percent Purity 98%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Citrinin, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0433163 Certificate TypeCertificate of Analysis Date19/02/2024 Catalog Number229580050
Lot NumberA0410386 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number229580050
compliance-icons
Product Identifier
  • Citrinin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Citrinin, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.