missing translation for 'onlineSavingsMsg'
Learn More
Citrinin, MP Biomedicals™ Product Code.: 11422072

Citrinin, MP Biomedicals™

Product Code. 11422072 Shop All MP Biomedicals Products
10 mg
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy

Product Code. 11422072

Brand: MP Biomedicals 0215070010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 518-75-2
Molecular Formula C13H14O5
Molecular Weight (g/mol) 250.25
InChI Key CBGDIJWINPWWJW-IYSWYEEDSA-N
Synonym citrinin, antimycin, --citrinin, citriain, 3r,4s-4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid, unii-3s697x6snz, ccris 175, 3h-2-benzopyran-7-carboxylic acid, 4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-, 3r-trans, 3r-trans-4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid, 4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acidShow MoreShow Less
PubChem CID 54680783
IUPAC Name (3R,4S)-6-hydroxy-3,4,5-trimethyl-8-oxo-3,4-dihydroisochromene-7-carboxylic acid
SMILES CC1C(OC=C2C1=C(C(=C(C2=O)C(=O)O)O)C)C

Specifications

Color Yellow
Quantity 10 mg
Formula Weight 250.3
Physical Form Crystalline Powder
Chemical Name or Material Citrinin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Citrinin, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.