missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CK 666, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Inhibitor of the Arp2/3 complex; inhibits actin polymerization (IC50 = 4 μM).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 296.34 |
Percent Purity | >99% |
Chemical Name or Material | CK 666 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'