Learn More
Thermo Scientific™ CL-XPosure™ Film
Thermo Scientific CL-XPosure Film is an economically priced clear-blue X-ray film especially for detection and documentation of ECL-based western blots and other chemiluminescent protein or nucleic acid assay methods.
Brand: Thermo Scientific™ 34090
146.37 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : PN
Additional Details : Weight : 0.10000kg
Description
Thermo Scientific CL-XPosure Film is an economically priced clear-blue X-ray film especially for detection and documentation of ECL-based western blots and other chemiluminescent protein or nucleic acid assay methods. CL-XPosure Film is an excellent photographic film for use with enhanced chemiluminescence (ECL) substrates for horseradish peroxidase (HRP) or alkaline phosphatase (AP). Convenient sizes are offered for protein western blotting, standard film cassettes, and film developing equipment. This autoradiography film also works well for chemiluminescent or isotopic DNA and RNA methods, including Southern and northern blotting and gel-shift assays (EMSA). The quality and price of this film make it the ideal choice for life science research applications.
Features include:
• High performance—just as sensitive as other commercially available autoradiography films
• Economical—achieve high-quality images at an affordable price
• Convenient—available in five sheet sizes in packages containing 100 non-interleaved sheets
• Perfect match—validated for use with SuperSignal chemiluminescent substrates and Pierce ECL and ECL Plus chemiluminescent substrates
Specifications
18 x 13 cm | |
CL-Xposure | |
7 in. | |
100 Sheets |
X-Ray Film | |
CL-XPosure Film, 7 x 5 in. (18 x 13 cm) | |
5 in. | |
Store at room temperature. |