Learn More
MedChemExpress Cladribine
MedChemExpress Cladribine (2-Chloro-2′-deoxyadenosine), a purine nucleoside analog, is an orally active adenosine deaminase inhibitor. Cladribine functions as an inhibitor of DNA synthesis to block the repair of the damaged DNA. Cladribine can inhibit DNA methylation. Cladribine has anti-lymphoma activity. Cladribine can be used for the research of several hematologic malignancies and multiple sclerosis.
Brand: MedChemExpress HY-13599
Specifications
285.69 g/mol | |
4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light) | |
98.92% | |
DMSO : ≥ 30 mg/mL (105.01 mM) ∣H2O : 10 mg/mL (35.00 mM∣Need ultrasonic) | |
4291-63-8 |
Reference compound | |
Room temperature | |
100 mg | |
COVID-19-immunoregulation | |
2-Chloro-2′-deoxyadenosine∣ CldAdo∣ 2CdA |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.