missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Clamp Connector

OHAUS™ Clamp Connector provides secure support for holding glassware at elevated positions.

Brand:  OHAUS™ 30392358

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15722409

  • £29.30 / Each
Estimated Shipment: 09-10-2024
to see stock.
Add to basket

Description

Description

  • Hold glassware safely
  • Corrosion-resistant
  • Providing convenience with exceptional stability and durability
Specifications

Specifications

Hook
Stainless Steel
Clamp, Connector, Hook, CLC-HOOKCS
Connector
Fixed 90° Connection Angle
0 to 13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers