Learn More
OHAUS™ Clamp Connector
OHAUS™ Clamp Connector provides secure support for holding glassware at elevated positions.
Brand: OHAUS™ 30392269
Additional Details : Weight : 0.50000kg
Description
- Hold glassware safely
- Corrosion-resistant
- Providing convenience with exceptional stability and durability
Specifications
Bar | |
Aluminum | |
203 mm length |
Connector | |
Clamp, Connector, Bar, CLC-HTZMBA20 |