Learn More
Fisherbrand™ Cellulose Stoppers
For flasks, erlenmeyers and test tubes.
Brand:  Fisherbrand™ 0005N00080
 View more versions of this product
Product Code. 11738533
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Permeable in air
  • Sterilizable to 200°C maximum
  • Single use
Specifications
Specifications
Single-use
60 mm
200°C
46 mm
Stopper
Beakers, Erlenmeyer flasks, tubes, and different types of bottles
100PK
35 mm
Cellulose Fiber
Sterile