Learn More
Kartell™ Clamps for Glass Joints
Plastic clamps for safe connection of standard-finish glass conical joints
£27.85 - £56.10
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15875577
|
Kartell™
3101900 |
10/19 |
£27.85
Pack of 10 |
Estimated Shipment: 22-11-2024
Log in to see stock. |
|||||
15825587
|
Kartell™
3343500 |
34/35 |
£52.30
Pack of 10 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
15835587
|
Kartell™
3454000 |
45/40 |
£56.10
Pack of 10 |
Estimated Shipment: 22-11-2024
Log in to see stock. |
|||||
Description
- Corrosion proof
- Do not scratch glass
- Available in various colours according to dimension
- Autoclavable
Specifications
10/Pk. | |
POM |