Learn More
Honeywell™ Class 1 Plastic Cartridge A1 for Organic Vapors
The A1 cartridge is part of the large choice of cartridges offered to protect against many hazardous gases and vapors in many applications. The range provides high quality and a cost-effective protection.
Brand: Honeywell™ N06575001L
Additional Details : Weight : 0.13500kg
Description
- Protects against organic vapors
- Excellent filtration efficiency over extensive period of time
- Lower profile and lightweight for optimal comfort
- Low profile does not obstruct the field of vision
- Threaded connector is easy to attach
Specifications
Class 1 plastic cartridge | |
Protects against organic vapors | |
73.9 g |
EC Category PPE: 3, QA: ISO 9001/2000, EC Certification: 2259, EC Attestation: 2259, EN 2259 | |
6 Pair/Box, 8 Boxes/Cs. |